2-Cyanobenzaldehyde

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Formylbenzonitrile

AS22056G100 AS22056G25
ACTIAS22056G100EA 0 GBP
ACTIAS22056G100 ACTIAS22056G25
2-Cyanobenzaldehyde
2-Cyanobenzaldehyde
Formula: C₈H₅NO
MW: 131.13 g/mol
MDL Number: MFCD00017503
CAS Number: 7468-67-9
UN: 3439
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR