2-Bromo-3-hydroxypyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

Synonyms: 2-Bromo-3-pyridinol

AS21514G100 AS21514G25
ACTIAS21514G100EA 0 GBP
ACTIAS21514G100 ACTIAS21514G25
2-Bromo-3-hydroxypyridine
2-Bromo-3-hydroxypyridine
Formula: C₅H₄BrNO
MW: 174 g/mol
Boiling Pt: 232
Melting Pt: 185…188 °C
Storage Temperature: Ambient
MDL Number: MFCD00006220
CAS Number: 6602-32-0
EINECS: 229-547-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR