2-Amino-3,5-dibromopyridine
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
AS21743G100
AS21743G25
ACTIAS21743G100EA
0
GBP
ACTIAS21743G100
ACTIAS21743G25
2-Amino-3,5-dibromopyridine
2-Amino-3,5-dibromopyridine
Formula:
C₅H₄Br₂N₂ Boiling Pt: 254 °C (1013 hPa) Melting Pt: 104 °C |
MDL Number:
MFCD00038041 CAS Number: 35486-42-1 EINECS: 252-590-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...