2-Bromo-3-fluorobenzoic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AS20663G1 AS20663G25 AS20663G5
ACTIAS20663G1EA 0 GBP
ACTIAS20663G1 ACTIAS20663G25 ACTIAS20663G5
2-Bromo-3-fluorobenzoic acid
2-Bromo-3-fluorobenzoic acid
Formula: C₇H₄BrFO₂
MDL Number: MFCD01569398
CAS Number: 132715-69-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR