4-Bromoanthranilic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Warning

AS22842G25 AS22842G5
ACTIAS22842G25EA 0 GBP
ACTIAS22842G25 ACTIAS22842G5
4-Bromoanthranilic acid
4-Bromoanthranilic acid
Formula: C₇H₆BrNO₂
MDL Number: MFCD03618454
CAS Number: 20776-50-5
EINECS: 244-025-7
UN: 2811
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR