trans-2-Bromocinnamic acid
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS24161G100
AS24161G25
ACTIAS24161G100EA
167
GBP
ACTIAS24161G100
ACTIAS24161G25
trans-2-Bromocinnamic acid
trans-2-Bromocinnamic acid
Formula:
C₉H₇BrO₂ MW: 227.06 g/mol |
MDL Number:
MFCD00016836 CAS Number: 7499-56-1 EINECS: 231-353-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...