2-Bromo-3-chloropyridine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS51096G100 AS51096G5
ACTIAS51096G100EA 0 GBP
ACTIAS51096G100 ACTIAS51096G5
2-Bromo-3-chloropyridine
2-Bromo-3-chloropyridine
Formula: C₅H₃BrClN
MDL Number: MFCD04039344
CAS Number: 96424-68-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR