2-Bromo-4-methylpyridine
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS20166G100
AS20166G25
ACTIAS20166G100EA
0
GBP
ACTIAS20166G100
ACTIAS20166G25
2-Bromo-4-methylpyridine
2-Bromo-4-methylpyridine
Formula:
C₆H₆BrN Boiling Pt: 42…44 °C (0.6 mmHg) Melting Pt: 218…228 °C Density: 1,545 g/cm³ (20 °C) |
MDL Number:
MFCD00082590 CAS Number: 4926-28-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...