Ethyl-6-(chloroformyl)hexanoate
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
011332-1G
011332-5G
FLUO011332-1GEA
0
GBP
FLUO011332-1G
FLUO011332-5G
Ethyl-6-(chloroformyl)hexanoate
Ethyl 6-(chloroformyl)hexanoate
Formula:
C₉H₁₅ClO₃ |
MDL Number:
MFCD00671507 CAS Number: 14794-32-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...