4'-(1-Pyrrolidinyl)acetophenone

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

OT-0677-5G OT-0677-100G
COBBOT-0677-5GEA 0 GBP
COBBOT-0677-5G COBBOT-0677-100G
4'-(1-Pyrrolidinyl)acetophenone
4'-(1-Pyrrolidinyl)acetophenone
Formula: C₁₂H₁₅NO
MW: 189.25 g/mol
MDL Number: MFCD01075691
CAS Number: 21557-09-5

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR