7-Fluoroquinaldine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

021590-5G 021590-25G
FLUO021590-5GEA 0 GBP
FLUO021590-5G FLUO021590-25G
7-Fluoroquinaldine
7-Fluoroquinaldine
Formula: C₁₀H₈FN
MDL Number: MFCD02683029
CAS Number: 1128-74-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR