7-Fluoroquinaldine

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BK-4504-100G BK-4504-5G BK-4504-25G
COBBBK-4504-100GEA 0 GBP
COBBBK-4504-100G COBBBK-4504-5G COBBBK-4504-25G
7-Fluoroquinaldine
7-Fluoroquinaldine
Formula: C₁₀H₈FN
MDL Number: MFCD02683029
CAS Number: 1128-74-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR