7-Fluoroquinaldine

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS25299G100 AS25299G25
ACTIAS25299G100EA 0 GBP
ACTIAS25299G100 ACTIAS25299G25
7-Fluoroquinaldine
7-Fluoroquinaldine
Formula: C₁₀H₈FN
MDL Number: MFCD02683029
CAS Number: 1128-74-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR