2,2,2,4'-Tetrafluoroacetophenone, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5


M-Clarity™ quality level = MQ200

402478-10G 402478-1G
SIAL402478-10GEA 218 GBP
SIAL402478-10G SIAL402478-1G
2,2,2,4'-Tetrafluoroacetophenone, Sigma-Aldrich®
2,2,2,4'-Tetrafluoroacetophenone
Formula: C₈H₄F₄O
MW: 192.1104 g/mol
MDL Number: MFCD00042393
CAS Number: 655-32-3
EINECS: 211-509-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR