2,2,2,4'-Tetrafluoroacetophenone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

002973-5G 002973-25G
FLUO002973-5GEA 0 GBP
FLUO002973-5G FLUO002973-25G
2,2,2,4'-Tetrafluoroacetophenone
2,2,2,4'-Tetrafluoroacetophenone
Formula: C₈H₄F₄O
MDL Number: MFCD00042393
CAS Number: 655-32-3
EINECS: 211-509-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR