α,4-Dibromo-3-fluorotoluene
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4-Bromo-3-fluorobenzyl bromide, 1-Bromo-4-(bromomethyl)-2-fluorobenzene
AS25475G25
AS25475G5
ACTIAS25475G25EA
0
GBP
ACTIAS25475G25
ACTIAS25475G5
α,4-Dibromo-3-fluorotoluene
α,4-Dibromo-3-fluorotoluene
Formula:
C₇H₅Br₂F |
MDL Number:
MFCD11847179 CAS Number: 127425-73-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...