α,2-Dibromo-5-fluorotoluene

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 1-Bromo-2-(bromomethyl)-4-fluorobenzene, 2-Bromo-5-fluorobenzyl bromide

AS24957G100 AS24957G25
ACTIAS24957G100EA 0 GBP
ACTIAS24957G100 ACTIAS24957G25
α,2-Dibromo-5-fluorotoluene
α,2-Dibromo-5-fluorotoluene
Formula: C₇H₅Br₂F
MW: 267.92 g/mol
MDL Number: MFCD00042187
CAS Number: 112399-50-5
UN: 3265
ADR: 8 II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR