4-tert-Butylphenylboronic acid pinacol ester

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-(4-(tert-Butyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane

021626-5G 021626-25G
FLUO021626-5GEA 0 GBP
FLUO021626-5G FLUO021626-25G
4-tert-Butylphenylboronic acid pinacol ester
4-tert-Butylphenylboronic acid pinacol ester
Formula: C₁₆H₂₅BO₂
MW: 260.1800 g/mol
MDL Number: MFCD05663843
CAS Number: 214360-66-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR