1,3-Dibromo-5-isopropylbenzene

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

023866-25G 023866-5G
FLUO023866-25GEA 0 GBP
FLUO023866-25G FLUO023866-5G
1,3-Dibromo-5-isopropylbenzene
1,3-Dibromo-5-isopropylbenzene
Formula: C₉H₁₀Br₂
MDL Number: MFCD06657957
CAS Number: 62655-20-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR