2-Fluoro-4-(trifluoromethyl)phenylboronic acid pinacol ester

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Fluoro-4-trifluoromethylphenylboronic acid pinacol ester, 2-(2-Fluoro-4-(trifluoromethyl)phenyl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 2-Fluoro-4-(trifluoromethyl)benzeneboronic acid pinacol ester

PN-3559-5G PN-3559-1G
COBBPN-3559-5GEA 0 GBP
COBBPN-3559-5G COBBPN-3559-1G
2-Fluoro-4-(trifluoromethyl)phenylboronic acid pinacol ester
2-Fluoro-4-(trifluoromethyl)phenylboronic acid pinacol ester
Formula: C₁₃H₁₅BF₄O₂
MW: 290.06 g/mol
MDL Number: MFCD08458187
CAS Number: 1073353-68-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR