N-Ethyl-3-fluorobenzylamine
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
N-(3-Fluorobenzyl)ethanamine
AS20443G1
AS20443G25
AS20443G5
ACTIAS20443G1EA
32
GBP
ACTIAS20443G1
ACTIAS20443G25
ACTIAS20443G5
N-Ethyl-3-fluorobenzylamine
N-Ethyl-3-fluorobenzylamine
Formula:
C₉H₁₂FN MW: 153.2000 g/mol |
MDL Number:
MFCD06655514 CAS Number: 90389-85-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...