3',5'-Dichloro-2'-methoxyacetophenone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

019223-25G 019223-5G
FLUO019223-25GEA 0 GBP
FLUO019223-25G FLUO019223-5G
3',5'-Dichloro-2'-methoxyacetophenone
3',5'-Dichloro-2'-methoxyacetophenone
Formula: C₉H₈Cl₂O₂
MDL Number: MFCD03789120
CAS Number: 154260-25-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR