(2,6-Difluoro-3-methylphenyl)acetonitrile
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
005765-5G
005765-10G
FLUO005765-5GEA
0
GBP
FLUO005765-5G
FLUO005765-10G
(2,6-Difluoro-3-methylphenyl)acetonitrile
(2,6-Difluoro-3-methylphenyl)acetonitrile
Formula:
C₉H₇F₂N |
MDL Number:
MFCD01631337 CAS Number: 261944-04-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...