(2,3-Difluoro-4-methylphenyl)acetonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

005764-1G 005764-2.5G
FLUO005764-1GEA 0 GBP
FLUO005764-1G FLUO005764-2.5G
(2,3-Difluoro-4-methylphenyl)acetonitrile
(2,3-Difluoro-4-methylphenyl)acetonitrile
Formula: C₉H₇F₂N
MDL Number: MFCD01631648
CAS Number: 261944-01-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR