4-(Difluoromethoxy)phenylacetonitrile

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

007872-1G 007872-5G
FLUO007872-1GEA 0 GBP
FLUO007872-1G FLUO007872-5G
4-(Difluoromethoxy)phenylacetonitrile
4-(Difluoromethoxy)phenylacetonitrile
Formula: C₉H₇F₂NO
MDL Number: MFCD00236242
CAS Number: 41429-16-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR