2'-Chloro-5'-nitroacetophenone
Supplier: ACTIVATE SCIENTIFIC
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
AS20530G25
AS20530G5
ACTIAS20530G25EA
0
GBP
ACTIAS20530G25
ACTIAS20530G5
2'-Chloro-5'-nitroacetophenone
2'-Chloro-5'-nitroacetophenone
Formula:
C₈H₆ClNO₃ MW: 199.5911 g/mol |
MDL Number:
MFCD00052836 CAS Number: 23082-50-0 EINECS: 245-421-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...