2-Methyl-4-(trifluoromethyl)phenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-3929-5G BB-3929-1G
COBBBB-3929-5GEA 0 GBP
COBBBB-3929-5G COBBBB-3929-1G
2-Methyl-4-(trifluoromethyl)phenylboronic acid
2-Methyl-4-(trifluoromethyl)phenylboronic acid
Formula: C₈H₈BF₃O₂
MDL Number: MFCD08689550
CAS Number: 957034-45-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR