5'-Fluoro-2'-methoxyacetophenone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

019293-1G 019293-5G
FLUO019293-1GEA 0 GBP
FLUO019293-1G FLUO019293-5G
5'-Fluoro-2'-methoxyacetophenone
5'-Fluoro-2'-methoxyacetophenone
Formula: C₉H₉FO₂
MDL Number: MFCD00671763
CAS Number: 445-82-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR