3'-Chloro-2,2,2,4'-tetrafluoroacetophenone
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
023135-1G
023135-5G
FLUO023135-1GEA
0
GBP
FLUO023135-1G
FLUO023135-5G
3'-Chloro-2,2,2,4'-tetrafluoroacetophenone
3'-Chloro-2,2,2,4'-tetrafluoroacetophenone
Formula:
C₈H₃ClF₄O MW: 226.555 g/mol |
MDL Number:
MFCD01319998 CAS Number: 845823-05-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...