ω-Chloro-3,4-difluoroacetophenone

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2-Chloro-3',4'-difluoroacetophenone, 2-Chloro-1-(3,4-difluorophenyl)ethanone

AS25280G100 AS25280G25 AS25280G5
ACTIAS25280G100EA 0 GBP
ACTIAS25280G100 ACTIAS25280G25 ACTIAS25280G5
ω-Chloro-3,4-difluoroacetophenone
ω-Chloro-3,4-difluoroacetophenone
Formula: C₈H₅ClF₂O
MDL Number: MFCD03966888
CAS Number: 51336-95-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR