Methyl perfluorodecanoate

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

002280-5G 002280-25G 002280-100G 002280-10G
FLUO002280-5GEA 0 GBP
FLUO002280-5G FLUO002280-25G FLUO002280-100G FLUO002280-10G
Methyl perfluorodecanoate
Methyl perfluorodecanoate
Formula: C₁₁H₃F₁₉O₂
MDL Number: MFCD00153211
CAS Number: 307-79-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR