3'-Bromo-4'-fluoroacetanilide

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS23607G1 AS23607G25 AS23607G5
ACTIAS23607G1EA 0 GBP
ACTIAS23607G1 ACTIAS23607G25 ACTIAS23607G5
3'-Bromo-4'-fluoroacetanilide
3'-Bromo-4'-fluoroacetanilide
Formula: C₈H₇BrFNO
MDL Number: MFCD18071051
CAS Number: 1009-75-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR