4-(Methylthio)phenol

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

003883-5G 003883-25G
FLUO003883-5GEA 0 GBP
FLUO003883-5G FLUO003883-25G
4-(Methylthio)phenol
4-(Methylthio)phenol
Formula: C₇H₈OS
MDL Number: MFCD00002351
CAS Number: 1073-72-9
EINECS: 214-031-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR