4-Fluoro-3-nitroanisole

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

AS32431G1 AS32431G5
ACTIAS32431G1EA 0 GBP
ACTIAS32431G1 ACTIAS32431G5
4-Fluoro-3-nitroanisole
4-Fluoro-3-nitroanisole
Formula: C₇H₆FNO₃
MDL Number: MFCD11848442
CAS Number: 61324-93-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR