4-(Difluoromethoxy)benzylamine

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: (4-(Difluoromethoxy)phenyl)methanamine

007860-5G 007860-1G
FLUO007860-5GEA 0 GBP
FLUO007860-5G FLUO007860-1G
4-(Difluoromethoxy)benzylamine
4-(Difluoromethoxy)benzylamine
Formula: C₈H₉F₂NO
MW: 173.16 g/mol
MDL Number: MFCD00236231
CAS Number: 177842-14-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR