4-Fluoro-3-methylbenzylamine
Supplier: FLUOROCHEM
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
005797-1G
005797-5G
FLUO005797-1GEA
0
GBP
FLUO005797-1G
FLUO005797-5G
4-Fluoro-3-methylbenzylamine
4-Fluoro-3-methylbenzylamine
Formula:
C₈H₁₀FN |
MDL Number:
MFCD01631490 CAS Number: 261951-68-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...