2,6-Difluoro-3-methylbenzamide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

005749-5G 005749-1G
FLUO005749-5GEA 0 GBP
FLUO005749-5G FLUO005749-1G
2,6-Difluoro-3-methylbenzamide
2,6-Difluoro-3-methylbenzamide
Formula: C₈H₇F₂NO
MDL Number: MFCD01631408
CAS Number: 261763-36-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR