2,3-Difluoro-4-methylbenzamide

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

005748-5G 005748-1G
FLUO005748-5GEA 0 GBP
FLUO005748-5G FLUO005748-1G
2,3-Difluoro-4-methylbenzamide
2,3-Difluoro-4-methylbenzamide
Formula: C₈H₇F₂NO
MDL Number: MFCD01631633
CAS Number: 261763-35-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR