4'-Fluoro-3'-(trifluoromethyl)acetophenone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

006379-1G 006379-5G 006379-25G
FLUO006379-1GEA 0 GBP
FLUO006379-1G FLUO006379-5G FLUO006379-25G
4'-Fluoro-3'-(trifluoromethyl)acetophenone
4'-Fluoro-3'-(trifluoromethyl)acetophenone
Formula: C₉H₆F₄O
MDL Number: MFCD00061252
CAS Number: 208173-24-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR