2'-Fluoro-5'-(trifluoromethyl)acetophenone

Supplier: FLUOROCHEM
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

006375-1G 006375-5G 006375-10G
FLUO006375-1GEA 0 GBP
FLUO006375-1G FLUO006375-5G FLUO006375-10G
2'-Fluoro-5'-(trifluoromethyl)acetophenone
2'-Fluoro-5'-(trifluoromethyl)acetophenone
Formula: C₉H₆F₄O
MDL Number: MFCD00061248
CAS Number: 202664-53-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR