4-Fluoro-2-methylphenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-Fluoro-2-methylbenzeneboronic acid, 2-Methyl-4-fluorophenylboronic acid

BB-2204-5G BB-2204-1G BB-2204-25G
COBBBB-2204-5GEA 0 GBP
COBBBB-2204-5G COBBBB-2204-1G COBBBB-2204-25G
4-Fluoro-2-methylphenylboronic acid
4-Fluoro-2-methylphenylboronic acid
Formula: C₇H₈BFO₂
MW: 153.95 g/mol
MDL Number: MFCD02093072
CAS Number: 139911-29-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR