2,4,6-Triisopropylphenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 2,4,6-Triisopropylbenzeneboronic acid, (2,4,6-Triisopropylphenyl)boronic acid

BB-8358-5G BB-8358-1G
COBBBB-8358-5GEA 0 GBP
COBBBB-8358-5G COBBBB-8358-1G
2,4,6-Triisopropylphenylboronic acid
2,4,6-Triisopropylphenylboronic acid
Formula: C₁₅H₂₅BO₂
MW: 248.16 g/mol
MDL Number: MFCD02683099
CAS Number: 154549-38-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR