4-Butylphenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-n-Butylbenzeneboronic acid

BB-8352-5G BB-8352-1G
COBBBB-8352-5GEA 0 GBP
COBBBB-8352-5G COBBBB-8352-1G
4-Butylphenylboronic acid
4-Butylphenylboronic acid
Formula: C₁₀H₁₅BO₂
MW: 178.04 g/mol
MDL Number: MFCD02093926
CAS Number: 145240-28-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR