4-Butylphenylboronic acid ≥98%
Supplier: Thermo Scientific
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
|
Warning
|
Synonyms:
4-n-Butylbenzeneboronic acid
L15584.03
L15584.06
L15584.03EA
37.9
GBP
L15584.03
L15584.06
4-Butylphenylboronic acid ≥98%
4-Butylphenylboronic acid
Purity: 98%
CAS No.: 145240-28-4
Molecular Formula: C10H15BO2
Formula Weight: 178.04
Melting Point: 103-106°
Beilstein Reference: 6920034
Formula:
C₁₀H₁₅BO₂ MW: 178.04 g/mol Melting Pt: 103…106 °C Storage Temperature: Ambient |
MDL Number:
MFCD02093926 CAS Number: 145240-28-4 |
Learn more
![](/m-uk.vwr.com/en_GB/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...