4-tert-Butylphenylboronic acid, Sigma-Aldrich®

Supplier: Merck
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 4-tert-Butylbenzeneboronic acid, (4-(tert-Butyl)phenyl)boronic acid

M-Clarity™ quality level = MQ100

480053-25G 480053-5G
SIAL480053-25GEA 428 GBP
SIAL480053-25G SIAL480053-5G
4-tert-Butylphenylboronic acid, Sigma-Aldrich®
4-tert-Butylphenylboronic acid
Formula: C₁₀H₁₅BO₂
MW: 178.04 g/mol
MDL Number: MFCD01009697
CAS Number: 123324-71-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR