4-tert-Butylphenylboronic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
4-tert-Butylbenzeneboronic acid, (4-(tert-Butyl)phenyl)boronic acid
BB-2280-5G
BB-2280-25G
BB-2280-100G
COBBBB-2280-5GEA
0
GBP
COBBBB-2280-5G
COBBBB-2280-25G
COBBBB-2280-100G
4-tert-Butylphenylboronic acid
4-tert-Butylphenylboronic acid
Formula:
C₁₀H₁₅BO₂ MW: 178.04 g/mol |
MDL Number:
MFCD01009697 CAS Number: 123324-71-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...