4-Isobutylphenylboronic acid

Supplier: COMBI-BLOCKS
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

BB-2441-100G BB-2441-5G BB-2441-1G BB-2441-25G
COBBBB-2441-100GEA 0 GBP
COBBBB-2441-100G COBBBB-2441-5G COBBBB-2441-1G COBBBB-2441-25G
4-Isobutylphenylboronic acid
4-Isobutylphenylboronic acid
Formula: C₁₀H₁₅BO₂
MDL Number: MFCD01632198
CAS Number: 153624-38-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR