4-sec-Butylphenylboronic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
BB-2134-5G
BB-2134-1G
COBBBB-2134-5GEA
0
GBP
COBBBB-2134-5G
COBBBB-2134-1G
4-sec-Butylphenylboronic acid
4-sec-Butylphenylboronic acid
Formula:
C₁₀H₁₅BO₂ |
MDL Number:
MFCD02179456 CAS Number: 850568-56-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...