3,4-Dimethoxycinnamic acid

Supplier: ACTIVATE SCIENTIFIC
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Synonyms: 3,4-Dimethoxycinnamic Acid

AS22681G100 AS22681G25
ACTIAS22681G100EA 0 GBP
ACTIAS22681G100 ACTIAS22681G25
3,4-Dimethoxycinnamic acid
3,4-Dimethoxycinnamic acid
Formula: C₁₁H₁₂O₄
MW: 208.21 g/mol
Boiling Pt: 343 °C (1013 hPa)
Melting Pt: 198…202 °C
MDL Number: MFCD00004387
CAS Number: 2316-26-9
EINECS: 219-025-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR