7-Fluoro-2-methylquinoline-8-boronic acid
Supplier: COMBI-BLOCKS
Ratings:
(No Reviews)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Synonyms:
(7-Fluoro-2-methylquinolin-8-yl)boronic acid
BB-3274-1G
BB-3274-5G
COBBBB-3274-1GEA
0
GBP
COBBBB-3274-1G
COBBBB-3274-5G
7-Fluoro-2-methylquinoline-8-boronic acid
7-Fluoro-2-methylquinoline-8-boronic acid
Formula:
C₁₀H₉BFNO₂ MW: 204.9900 g/mol |
MDL Number:
MFCD11504838 CAS Number: 1072945-61-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...